Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cnppd1
Entrez Gene ID
69171
Gene Name
cyclin Pas1/PHO80 domain containing 1
Gene Aliases
1810031K17Rik, AU043960
Chromosome Location
Chr.1: 75135215 - 75142733 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026977.2 NP_081253.1 1 - 2 315 59
GenBank mRNA AK029090.1 - 1 - 2 315 59
AK145962.1 - 2 - 3 562 59
AK169779.1 - 1 - 2 587 59
BC028815.1 - 1 - 2 300 59

View Full Product Details