The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288612.1 | NP_001275541.1 | 1 - 2 | 185 | 64 |
NR_109938.1 | 1 - 2 | 185 | 64 | ||
GenBank mRNA | BC079884.1 | - | 1 - 2 | 130 | 64 |