Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Gulp1
- Entrez Gene ID
-
70676
- Gene Name
- GULP, engulfment adaptor PTB domain containing 1
- Gene Aliases
-
3110030A04Rik, 5730529O06Rik, CED-6, Ced6, GULP, Gulp-2
- Chromosome Location
-
Chr.1: 44551496 - 44796838 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK028490.1 |
- |
2 - 3 |
255 |
69 |
View Full Product Details