The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252458.1 | NP_001239387.1 | 15 - 16 | 2247 | 66 |
NM_178900.4 | NP_849231.1 | 14 - 15 | 2532 | 66 | |
GenBank mRNA | AK037030.1 | - | 14 - 15 | 2533 | 66 |
AK154029.1 | - | 14 - 15 | 2381 | 66 | |
BC095949.1 | - | 15 - 16 | 2197 | 66 | |
BC096444.1 | - | 15 - 16 | 2215 | 66 |