Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Synpo
- Entrez Gene ID
-
104027
- Gene Name
- synaptopodin
- Gene Aliases
-
9030217H17Rik, 9130229N11, 9330140I15Rik, AW046661
- Chromosome Location
-
Chr.18: 60593981 - 60660174 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK033711.1 |
- |
3 - 4 |
786 |
67 |
View Full Product Details