The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204907.1 | NP_001191836.1 | 15 - 16 | 1959 | 75 |
GenBank mRNA | AB017105.1 | - | 14 - 15 | 75 | |
AK138325.1 | - | 15 - 16 | 1915 | 75 |