Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Skint2
- Entrez Gene ID
-
329919
- Gene Name
- selection and upkeep of intraepithelial T cells 2
- Gene Aliases
-
A430070M19, B7S3, OTTMUSG00000008540, Skint-2
- Chromosome Location
-
Chr.4: 112613609 - 112652248 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC042786.1 |
- |
2 - 3 |
265 |
60 |
EF494893.1 |
- |
1 - 2 |
98 |
60 |
View Full Product Details