The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285463.1 | NP_001272392.1 | 17 - 18 | 2308 | 107 |
NM_001285473.1 | NP_001272402.1 | 13 - 14 | 1898 | 107 | |
NM_139150.5 | NP_631889.1 | 15 - 16 | 2181 | 107 | |
NR_104344.1 | 11 - 12 | 1633 | 107 | ||
GenBank mRNA | AB221600.1 | - | 14 - 15 | 1964 | 107 |
AF454947.1 | - | 15 - 16 | 2150 | 107 | |
AK044738.1 | - | 17 - 18 | 2309 | 107 | |
BC138644.1 | - | 14 - 15 | 2087 | 107 | |
BC138645.1 | - | 14 - 15 | 2087 | 107 | |
BC145350.1 | - | 14 - 15 | 2077 | 107 | |
EU170502.1 | - | 13 - 14 | 1879 | 107 | |
EU170503.1 | - | 11 - 12 | 1614 | 107 |