The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190371.1 | NP_001177300.1 | 9 - 10 | 909 | 99 |
GenBank mRNA | AK090336.1 | - | 9 - 10 | 862 | 99 |
AK162301.1 | - | 4 - 5 | 337 | 99 |