The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019584.3 | NP_062530.2 | 11 - 12 | 1310 | 90 |
GenBank mRNA | AF077302.2 | - | 10 - 11 | 1172 | 90 |
AK028851.1 | - | 9 - 10 | 2752 | 90 | |
AK031501.1 | - | 10 - 11 | 1937 | 90 | |
AK032176.1 | - | - | 1311 | 90 | |
AK050329.1 | - | 11 - 12 | 1311 | 90 | |
AK077392.1 | - | 11 - 12 | 1397 | 90 | |
AK083800.1 | - | 11 - 12 | 1300 | 90 | |
AK151531.1 | - | 11 - 12 | 1277 | 90 | |
BC005770.1 | - | 11 - 12 | 1291 | 90 |