Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
L1td1
Entrez Gene ID
381591
Gene Name
LINE-1 type transposase domain containing 1
Gene Aliases
AA546746, AB211064, D76865, ECAT11
Chromosome Location
Chr.4: 98726754 - 98738486 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081202.1 NP_001074671.1 2 - 3 163 93
GenBank mRNA AB211064.1 - 2 - 3 169 93
AK145814.1 - 2 - 3 156 93

View Full Product Details