Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rbm8a
Entrez Gene ID
60365
Gene Name
RNA binding motif protein 8a
Gene Aliases
2310057C03Rik, AA673428, Rbm8
Chromosome Location
Chr.3: 96629928 - 96633791 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102407.1 NP_001095877.1 1 - 2 106 75
NM_025875.2 NP_080151.2 1 - 2 106 75
GenBank mRNA AK010284.1 - 1 - 2 94 75
AK082925.1 - 1 - 2 95 75
BC020086.1 - 1 - 2 89 75
BC058376.1 - 1 - 2 78 75
BY128307.1 - 1 - 2 103 75
CJ072219.1 - 1 - 2 107 75
CJ075560.1 - 1 - 2 107 75
KF051021.1 - 1 - 2 72 75

View Full Product Details