Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nudt17
Entrez Gene ID
78373
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 17
Gene Aliases
2410015C20Rik
Chromosome Location
Chr.3: 96705892 - 96708929 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162925.1 NP_001156397.1 1 - 2 230 80
NM_030094.1 NP_084370.1 1 - 2 230 80
GenBank mRNA AK019109.1 - 1 - 2 235 80
AK134929.1 - 1 - 2 218 80
AK141256.1 - 1 - 2 218 80
BC115559.2 - 1 - 2 215 80

View Full Product Details