The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163170.1 | NP_001156642.1 | 5 - 6 | 818 | 68 |
GenBank mRNA | AK051276.1 | - | 5 - 6 | 803 | 68 |