The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008121.3 | NP_032147.1 | 1 - 2 | 85 | 93 |
GenBank mRNA | BC053054.1 | - | 1 - 2 | 71 | 93 |