Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd35
Entrez Gene ID
213121
Gene Name
ankyrin repeat domain 35
Gene Aliases
4732436F15Rik
Chromosome Location
Chr.3: 96670131 - 96691034 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081139.1 NP_001074608.1 10 - 11 3050 86
GenBank mRNA AK028691.1 - 10 - 11 2840 86
AK046624.1 - 10 - 11 2860 86
BC138276.1 - 10 - 11 2855 86
BC145228.1 - 10 - 11 2859 86
BC145750.1 - 10 - 11 2855 86
BC157973.1 - 10 - 11 2855 86

View Full Product Details