The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015732.4 | NP_056547.3 | 10 - 11 | 2741 | 65 |
GenBank mRNA | AF073788.1 | - | 10 - 11 | 2606 | 65 |
AF205889.1 | - | 9 - 10 | 2446 | 65 | |
AK049192.1 | - | 10 - 11 | 2686 | 65 | |
AK084644.1 | - | 3 - 4 | 526 | 65 | |
AK198476.1 | - | 2 - 3 | 262 | 65 | |
AK202759.1 | - | 2 - 3 | 262 | 65 | |
BC057338.1 | - | 10 - 11 | 2724 | 65 |