The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026360.3 | NP_080636.2 | 4 - 5 | 478 | 100 |
GenBank mRNA | AK010310.1 | - | 4 - 5 | 471 | 100 |
AK076982.1 | - | 4 - 5 | 463 | 100 | |
AK169595.1 | - | 2 - 3 | 101 | 100 | |
BC060168.1 | - | 4 - 5 | 458 | 100 | |
BC095944.1 | - | 4 - 5 | 455 | 100 | |
BC132242.1 | - | 4 - 5 | 470 | 100 | |
BC132244.1 | - | 4 - 5 | 470 | 100 |