The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026360.3 | NP_080636.2 | 9 - 10 | 1062 | 84 |
GenBank mRNA | AK010310.1 | - | 9 - 10 | 1055 | 84 |
AK076982.1 | - | 9 - 10 | 1047 | 84 | |
AK169595.1 | - | 7 - 8 | 685 | 84 | |
BC006843.1 | - | 2 - 3 | 249 | 84 | |
BC095944.1 | - | 9 - 10 | 1039 | 84 | |
BC132242.1 | - | 9 - 10 | 1054 | 84 | |
BC132244.1 | - | 9 - 10 | 1054 | 84 |