The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178280.3 | NP_840064.2 | 3 - 4 | 3355 | 67 |
GenBank mRNA | AK045051.1 | - | 2 - 3 | 1241 | 67 |
BC072631.1 | - | 3 - 4 | 3299 | 67 | |
BC094218.1 | - | 2 - 3 | 543 | 67 | |
BC118974.1 | - | 2 - 3 | 3633 | 67 | |
BC128071.1 | - | 2 - 3 | 3632 | 67 | |
X97581.1 | - | 1 - 2 | 3443 | 67 |