The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146188.1 | NP_666300.1 | 1 - 2 | 261 | 74 |
GenBank mRNA | AK034731.1 | - | 2 - 3 | 333 | 74 |
BC031749.1 | - | 1 - 2 | 261 | 74 |