The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017394.4 | NP_059090.3 | 10 - 11 | 1556 | 113 |
GenBank mRNA | AB026688.1 | - | 10 - 11 | 1470 | 113 |
AK005282.1 | - | 8 - 9 | 953 | 113 | |
AK031867.1 | - | 10 - 11 | 1524 | 113 | |
BC054765.1 | - | 10 - 11 | 1499 | 113 |