The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175028.1 | NP_778193.1 | 1 - 2 | 118 | 110 |
GenBank mRNA | AB093268.1 | - | 1 - 2 | 118 | 110 |