The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175028.1 | NP_778193.1 | 3 - 4 | 326 | 69 |
GenBank mRNA | AB093268.1 | - | 3 - 4 | 326 | 69 |
BC044898.1 | - | 3 - 4 | 231 | 69 | |
BC047352.1 | - | 3 - 4 | 231 | 69 | |
BC064672.1 | - | 1 - 2 | 99 | 69 |