The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172632.2 | NP_766220.2 | 5 - 6 | 2001 | 80 |
GenBank mRNA | AK079654.1 | - | 5 - 6 | 1979 | 80 |
BC058942.1 | - | 4 - 5 | 1404 | 80 | |
BC062911.1 | - | 4 - 5 | 1404 | 80 | |
BC094564.1 | - | 4 - 5 | 2215 | 80 |