Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nol9
Entrez Gene ID
74035
Gene Name
nucleolar protein 9
Gene Aliases
4632412I24Rik, 6030462G04Rik, AI449622, AW490720
Chromosome Location
Chr.4: 152039274 - 152061494 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK014568.1 - 9 - 10 1718 67
AK042581.1 - 9 - 10 1704 67
AK157398.1 - 9 - 10 1716 67
AK157956.1 - 10 - 11 1621 67
AK164493.1 - 9 - 10 1727 67
AK172043.1 - 9 - 10 1722 67
BC051109.1 - 8 - 9 1601 67
BC132099.1 - 9 - 10 1715 67

View Full Product Details