Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnfrsf14
Entrez Gene ID
230979
Gene Name
tumor necrosis factor receptor superfamily, member 14 (herpesvirus entry mediator)
Gene Aliases
Atar, HveA, Hvem, Tnfrs14
Chromosome Location
Chr.4: 154921442 - 154928620 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF515707.1 - 7 - 8 751 69
BC022125.1 - 5 - 6 513 69
BC104055.1 - 7 - 8 782 69

View Full Product Details