Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ski
Entrez Gene ID
20481
Gene Name
ski sarcoma viral oncogene homolog (avian)
Gene Aliases
2310012I02Rik, 2610001A11Rik, AA062172, AA589460, BC004088
Chromosome Location
Chr.4: 155154075 - 155222593 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011385.2 NP_035515.2 5 - 6 1781 102
GenBank mRNA AF435852.1 - 5 - 6 102
AK019148.2 - 5 - 6 838 102
BC030841.1 - 2 - 3 558 102
BC068305.1 - 6 - 7 1625 102
CB722801.1 - 2 - 3 490 102

View Full Product Details