The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113360.2 | NP_001106831.1 | 2 - 3 | 486 | 62 |
NM_175556.4 | NP_780765.2 | 2 - 3 | 486 | 62 | |
GenBank mRNA | AK044619.1 | - | 2 - 3 | 485 | 62 |
AK078731.1 | - | 2 - 3 | 257 | 62 | |
AK129147.1 | - | 2 - 3 | 186 | 62 | |
AY966876.1 | - | 2 - 3 | 264 | 62 | |
DQ176851.1 | - | 2 - 3 | 486 | 62 |