Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bnc2
Entrez Gene ID
242509
Gene Name
basonuclin 2
Gene Aliases
5031434M05Rik, 8430420F16Rik
Chromosome Location
Chr.4: 84266804 - 84676342 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK031440.1 - 3 - 4 244 96

View Full Product Details