Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mettl2
Entrez Gene ID
52686
Gene Name
methyltransferase like 2
Gene Aliases
2810438F06Rik, AW493404, C130031G21Rik, D11Ertd768e, METL, PSENIP1
Chromosome Location
Chr.11: 105126425 - 105141146 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172567.3 NP_766155.3 2 - 3 270 137
GenBank mRNA AK048040.1 - 2 - 3 271 137
AK153431.1 - 2 - 3 204 137
BC089591.1 - 2 - 3 208 137

View Full Product Details