Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cep76
Entrez Gene ID
225659
Gene Name
centrosomal protein 76
Gene Aliases
6230425F05Rik, 9630013E05
Chromosome Location
Chr.18: 67617397 - 67641336 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081073.1 NP_001074542.1 11 - 12 2002 104
GenBank mRNA AK035879.1 - 10 - 11 1782 104
AK078034.1 - 1 - 2 146 104
BC119216.1 - 11 - 12 1883 104
BC119218.1 - 11 - 12 1883 104

View Full Product Details