Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cep76
Entrez Gene ID
225659
Gene Name
centrosomal protein 76
Gene Aliases
6230425F05Rik, 9630013E05
Chromosome Location
Chr.18: 67617397 - 67641336 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081073.1 NP_001074542.1 5 - 6 867 91
GenBank mRNA AK035879.1 - 4 - 5 647 91
BC119216.1 - 5 - 6 748 91
BC119218.1 - 5 - 6 748 91

View Full Product Details