Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim32
Entrez Gene ID
69807
Gene Name
tripartite motif-containing 32
Gene Aliases
1810045E12Rik, 3f3, BBS11, Zfp117
Chromosome Location
Chr.4: 65604986 - 65616240 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161782.1 NP_001155254.1 1 - 2 83 59
GenBank mRNA AX775777.1 - 1 - 2 66 59

View Full Product Details