Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rergl
Entrez Gene ID
632971
Gene Name
RERG/RAS-like
Gene Aliases
EG632971
Chromosome Location
Chr.6: 139493182 - 139501909 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128090.1 NP_001121562.1 4 - 5 365 95
GenBank mRNA BC147153.1 - 4 - 5 365 95
BC147154.1 - 4 - 5 365 95
BC147732.1 - 4 - 5 430 95
BC147735.1 - 4 - 5 440 95

View Full Product Details