Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wnk3
Entrez Gene ID
279561
Gene Name
WNK lysine deficient protein kinase 3
Gene Aliases
Prkwnk3, Wnk3-ps
Chromosome Location
Chr.X: 151168777 - 151320194 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271678.1 NP_001258607.1 19 - 20 4596 109
NM_001271679.1 NP_001258608.1 19 - 20 4455 109
GenBank mRNA BC043119.1 - 2 - 3 649 109
BC060731.1 - 2 - 3 649 109
JQ247189.1 - 19 - 20 4596 109
JQ247190.1 - 19 - 20 4455 109

View Full Product Details