The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271678.1 | NP_001258607.1 | 21 - 22 | 4999 | 94 |
NM_001271679.1 | NP_001258608.1 | 21 - 22 | 4858 | 94 | |
GenBank mRNA | BC043119.1 | - | 4 - 5 | 1052 | 94 |
BC060731.1 | - | 4 - 5 | 1052 | 94 | |
JQ247189.1 | - | 21 - 22 | 4999 | 94 | |
JQ247190.1 | - | 21 - 22 | 4858 | 94 |