Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Whrn
Entrez Gene ID
73750
Gene Name
whirlin
Gene Aliases
1110035G07Rik, AW122018, AW742671, C430046P22Rik, Dfnb31, wi
Chromosome Location
Chr.4: 63414909 - 63496078 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008791.2 NP_001008791.1 4 - 5 1929 89
NM_001008793.2 NP_001008793.1 4 - 5 1929 89
NM_028640.2 NP_082916.1 4 - 5 1929 89
GenBank mRNA AK122523.1 - 4 - 5 1060 89
AK157955.1 - 4 - 5 1930 89
AY739114.1 - 4 - 5 1889 89
AY739115.1 - 4 - 5 1889 89
AY739117.1 - 4 - 5 1889 89
BC150757.1 - 4 - 5 1286 89
HQ148552.1 - 4 - 5 1171 89

View Full Product Details