Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mllt3
Entrez Gene ID
70122
Gene Name
myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 3
Gene Aliases
2210011H10Rik, 2610012I03Rik, 3830408D16Rik, Af9, D4Ertd321e
Chromosome Location
Chr.4: 87769925 - 88033407 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027326.3 NP_081602.3 10 - 11 1888 107
NM_029931.3 NP_084207.2 6 - 7 561 107
GenBank mRNA AF333960.1 - 9 - 10 1549 107
AK008707.1 - 6 - 7 523 107
AK147009.1 - 6 - 7 475 107
BC021420.1 - 11 - 12 1727 107

View Full Product Details