The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026983.2 | NP_081259.1 | 2 - 3 | 71 | |
GenBank mRNA | AK007684.1 | - | 2 - 3 | 71 | |
AK131830.1 | - | 2 - 3 | 71 | ||
BC037376.1 | - | 1 - 2 | 71 | ||
BC104406.1 | - | 2 - 3 | 234 | 71 | |
BC104407.2 | - | 2 - 3 | 234 | 71 |