Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sh3yl1
Entrez Gene ID
24057
Gene Name
Sh3 domain YSC-like 1
Gene Aliases
AI314953, Ray, YSC84
Chromosome Location
Chr.12: 30911669 - 30960160 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289480.1 NP_001276409.1 1 - 2 557 103
NM_013709.5 NP_038737.1 1 - 2 557 103
GenBank mRNA AK141199.1 - 1 - 2 558 103
BC058557.1 - 1 - 2 534 103
BC061488.1 - 1 - 2 497 103
BC110043.1 - 1 - 2 535 103

View Full Product Details