The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289480.1 | NP_001276409.1 | 8 - 9 | 1276 | 125 |
NM_013709.5 | NP_038737.1 | 9 - 10 | 1390 | 125 | |
NR_110343.1 | 10 - 11 | 1535 | 125 | ||
GenBank mRNA | AK141199.1 | - | 9 - 10 | 1391 | 125 |
AK168483.1 | - | 10 - 11 | 1016 | 125 | |
BC010296.1 | - | 8 - 9 | 838 | 125 | |
BC058557.1 | - | 8 - 9 | 1253 | 125 | |
BC061488.1 | - | 8 - 9 | 1990 | 125 | |
BC110043.1 | - | 9 - 10 | 1368 | 125 | |
D85926.1 | - | 9 - 10 | 873 | 125 |