The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198631.2 | NP_941033.2 | 11 - 12 | 1747 | 133 |
GenBank mRNA | AK129275.1 | - | 6 - 7 | 871 | 133 |