Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc9
Entrez Gene ID
243846
Gene Name
coiled-coil domain containing 9
Gene Aliases
2600011L02Rik
Chromosome Location
Chr.7: 16273534 - 16286815 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136471.1 NP_001129943.1 2 - 3 146 93
NM_172297.1 NP_758501.2 1 - 2 163 93
GenBank mRNA AK134892.1 - 2 - 3 132 93
BC021915.1 - 1 - 2 163 93

View Full Product Details