Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Strn4
Entrez Gene ID
97387
Gene Name
striatin, calmodulin binding protein 4
Gene Aliases
C80611, ZIN, zinedin
Chromosome Location
Chr.7: 16815889 - 16840931 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039878.2 NP_001034967.1 2 - 3 846 121
NM_133789.3 NP_598550.2 2 - 3 846 121
GenBank mRNA AK136155.1 - 2 - 3 162 121
AK154615.1 - 2 - 3 396 121
AK161673.1 - 2 - 3 418 121
BC080283.1 - 2 - 3 412 121

View Full Product Details