Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cndp2
Entrez Gene ID
66054
Gene Name
CNDP dipeptidase 2 (metallopeptidase M20 family)
Gene Aliases
0610010E05Rik, C76600, Cn2, Dip-2, Pep-1, Pep1
Chromosome Location
Chr.18: 84667465 - 84685702 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289531.1 NP_001276460.1 1 - 2 223 91
NM_023149.3 NP_075638.2 2 - 3 191 91
GenBank mRNA AB046738.1 - 2 - 3 132 91
AK003779.1 - 2 - 3 112 91
AK131124.1 - 1 - 2 103 91
AK149751.1 - 1 - 2 147 91
AK151013.1 - 2 - 3 113 91
AK152442.1 - 1 - 2 189 91
AK152729.1 - 1 - 2 161 91
AK153040.1 - 1 - 2 161 91
AK159129.1 - 1 - 2 189 91
AK159739.1 - 2 - 3 119 91
AK168029.1 - 2 - 3 130 91
AK168652.1 - 2 - 3 142 91
AK172034.1 - 2 - 3 113 91
BC005532.1 - 2 - 3 122 91
CJ078789.1 - 2 - 3 191 91
CN721222.1 - 1 - 2 223 91

View Full Product Details