The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026647.3 | NP_080923.2 | 3 - 4 | 519 | 103 |
GenBank mRNA | AK020293.1 | - | 3 - 4 | 401 | 103 |
AK144108.1 | - | 3 - 4 | 418 | 103 | |
AK148344.1 | - | 3 - 4 | 412 | 103 | |
AY166674.1 | - | 1 - 2 | 157 | 103 | |
BC043110.1 | - | 3 - 4 | 365 | 103 | |
BC058372.1 | - | 3 - 4 | 387 | 103 |