The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010720.3 | NP_034850.3 | 9 - 10 | 1775 | 81 |
GenBank mRNA | AF118768.1 | - | 9 - 10 | 1760 | 81 |
AK090224.1 | - | 9 - 10 | 1775 | 81 | |
AK150054.1 | - | 2 - 3 | 158 | 81 | |
AK150061.1 | - | 2 - 3 | 158 | 81 | |
AK152430.1 | - | 9 - 10 | 1773 | 81 | |
AK172101.1 | - | 9 - 10 | 1775 | 81 | |
BC020991.1 | - | 9 - 10 | 1629 | 81 |