Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stam2
Entrez Gene ID
56324
Gene Name
signal transducing adaptor molecule (SH3 domain and ITAM motif) 2
Gene Aliases
1200004O12Rik, 5730456G07Rik, BB145015, Hbp
Chromosome Location
Chr.2: 52692198 - 52742149 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019667.2 NP_062641.1 1 - 2 92 65
GenBank mRNA AB012611.1 - 1 - 2 107 65
AK004604.2 - 1 - 2 92 65
AK035789.1 - - 144 65
AK044230.1 - 1 - 2 120 65
AK078244.1 - 1 - 2 124 65
AK159699.1 - 1 - 2 121 65
AK168729.1 - 1 - 2 124 65
BC013818.1 - 1 - 2 128 65

View Full Product Details