The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285991.1 | NP_001272920.1 | 11 - 12 | 1101 | 70 |
NM_001285993.1 | NP_001272922.1 | 11 - 12 | 1068 | 70 | |
NM_028162.4 | NP_082438.3 | 11 - 12 | 1101 | 70 | |
GenBank mRNA | AK005461.1 | - | 11 - 12 | 1085 | 70 |
AK033710.1 | - | 11 - 12 | 993 | 70 | |
AK144467.1 | - | 11 - 12 | 1102 | 70 | |
AK154975.1 | - | 11 - 12 | 1061 | 70 | |
AK163366.1 | - | 11 - 12 | 1107 | 70 | |
AK220251.1 | - | 11 - 12 | 1068 | 70 | |
BC043113.1 | - | 11 - 12 | 917 | 70 |